Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCTTGAAGAAAAGAAGCCGCTGG[C/T]GCTCGGCGGCCTGGCAGGTCTCAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606908 MIM: 606513 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARFGAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARFGAP2 - ADP ribosylation factor GTPase activating protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6745 - microRNA 6745 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PACSIN3 - protein kinase C and casein kinase substrate in neurons 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184974.1 | 921 | Missense Mutation | CAC,CGC | H,R 236 | NP_001171903.1 | |
NM_001184975.1 | 921 | Missense Mutation | CAC,CGC | H,R 236 | NP_001171904.1 | |
NM_016223.4 | 921 | Missense Mutation | CAC,CGC | H,R 236 | NP_057307.2 |