Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCAGGAGATAGAAGCAGGTCTC[A/G]TACAAATTCCCATAGATGAGAACTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 115500 MIM: 605169 | ||||||||||||||||||||
Literature Links: |
CAT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAT - catalase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ELF5 - E74 like ETS transcription factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243080.1 | 532 | Nonsense Mutation | CGA,TGA | R,* 75 | NP_001230009.1 | |
NM_001243081.1 | 532 | Nonsense Mutation | CGA,TGA | R,* 102 | NP_001230010.1 | |
NM_001422.3 | 532 | Nonsense Mutation | CGA,TGA | R,* 170 | NP_001413.1 | |
NM_198381.1 | 532 | Nonsense Mutation | CGA,TGA | R,* 180 | NP_938195.1 | |
XM_017017308.1 | 532 | Nonsense Mutation | CGA,TGA | R,* 153 | XP_016872797.1 | |
XM_017017309.1 | 532 | Nonsense Mutation | CGA,TGA | R,* 147 | XP_016872798.1 |