Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGAGCCCCCTCGCATTCGCGGAG[C/T]CGGGACACGAGGTGGGTGCCTCCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 118445 | ||||||||||||||||||||
Literature Links: |
CCKBR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCKBR - cholecystokinin B receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318029.1 | 458 | Missense Mutation | GCC,GTC | A,V 47 | NP_001304958.1 | |
NM_176875.3 | 458 | Missense Mutation | GCC,GTC | A,V 47 | NP_795344.1 | |
XM_005253210.1 | 458 | Missense Mutation | GCC,GTC | A,V 47 | XP_005253267.1 | |
XM_017018516.1 | 458 | Intron | XP_016874005.1 |