Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTGGGGGCTGCCGTGTTTGGCA[C/G]CTTGGCAGACAGGTGAGCCCGTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 114220 MIM: 611696 | ||||||||||||||||||||
Literature Links: |
CAPN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAPN1 - calpain 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC22A20 - solute carrier family 22 member 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004326.4 | 506 | Missense Mutation | ACC,AGC | T,S 158 | NP_001004326.4 | |
XM_011545043.2 | 506 | Missense Mutation | ACC,AGC | T,S 158 | XP_011543345.1 | |
XM_017017774.1 | 506 | Missense Mutation | ACC,AGC | T,S 158 | XP_016873263.1 | |
XM_017017775.1 | 506 | Intron | XP_016873264.1 | |||
XM_017017776.1 | 506 | Intron | XP_016873265.1 |