Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCTGGGTGATCATGCCCACCCA[G/T]ATGTTAGGGAAGGCTGTGGTGTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608752 MIM: 606227 MIM: 606130 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QTNF5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1QTNF5 - C1q and tumor necrosis factor related protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278431.1 | 1621 | Intron | NP_001265360.1 | |||
NM_015645.4 | 1621 | UTR 5 | NP_056460.1 |
MFRP - membrane frizzled-related protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031433.3 | 1621 | Silent Mutation | NP_113621.1 |
RNF26 - ring finger protein 26 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |