Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCATTAATGCGGGGGACAGAAGA[A/T]GCCGAGTCTCGACTGCCCATGTCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613306 | ||||||||||||||||||||
Literature Links: |
ALKBH8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALKBH8 - alkB homolog 8, tRNA methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301010.1 | 1577 | Silent Mutation | GCA,GCT | A,A 557 | NP_001287939.1 | |
NM_138775.2 | 1577 | Silent Mutation | GCA,GCT | A,A 554 | NP_620130.2 | |
XM_005271728.2 | 1577 | Silent Mutation | GCA,GCT | A,A 417 | XP_005271785.1 | |
XM_005271729.3 | 1577 | Silent Mutation | GCA,GCT | A,A 417 | XP_005271786.1 | |
XM_017018553.1 | 1577 | Silent Mutation | GCA,GCT | A,A 507 | XP_016874042.1 | |
XM_017018554.1 | 1577 | Silent Mutation | GCA,GCT | A,A 504 | XP_016874043.1 | |
XM_017018555.1 | 1577 | Silent Mutation | GCA,GCT | A,A 302 | XP_016874044.1 | |
XM_017018556.1 | 1577 | Silent Mutation | GCA,GCT | A,A 252 | XP_016874045.1 | |
XM_017018557.1 | 1577 | Intron | XP_016874046.1 |