Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCAAAATTCCTGTTTTATAGTAG[C/T]ATCTGAAAAAGCAAGCAGAAGAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 115500 MIM: 605169 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAT - catalase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ELF5 - E74 like ETS transcription factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243080.1 | 698 | Missense Mutation | TAC,TGC | Y,C 130 | NP_001230009.1 | |
NM_001243081.1 | 698 | Missense Mutation | TAC,TGC | Y,C 157 | NP_001230010.1 | |
NM_001422.3 | 698 | Missense Mutation | TAC,TGC | Y,C 225 | NP_001413.1 | |
NM_198381.1 | 698 | Missense Mutation | TAC,TGC | Y,C 235 | NP_938195.1 | |
XM_017017308.1 | 698 | Missense Mutation | TAC,TGC | Y,C 208 | XP_016872797.1 | |
XM_017017309.1 | 698 | Missense Mutation | TAC,TGC | Y,C 202 | XP_016872798.1 |