Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTTGAGAGCAAAATGACAACACC[A/C]AGAAATTCAGTAAATGGGACTTTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 112210 | ||||||||||||||||||||
Literature Links: |
MS4A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MS4A1 - membrane spanning 4-domains A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021950.3 | 165 | Silent Mutation | CCA,CCC | P,P 4 | NP_068769.2 | |
NM_152866.2 | 165 | Silent Mutation | CCA,CCC | P,P 4 | NP_690605.1 |