Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACTCGAGTTACCCCAAGATCGAT[A/G]AGGTCCTCCTTCTGCAGGTTTGGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603290 | ||||||||||||||||||||
Literature Links: |
SHANK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SHANK2 - SH3 and multiple ankyrin repeat domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012309.4 | 6825 | Silent Mutation | CTC,CTT | L,L 1825 | NP_036441.2 | |
NM_133266.4 | 6825 | Silent Mutation | CTC,CTT | L,L 1237 | NP_573573.2 | |
XM_005277932.3 | 6825 | Silent Mutation | CTC,CTT | L,L 1446 | XP_005277989.1 | |
XM_017017387.1 | 6825 | Silent Mutation | CTC,CTT | L,L 1825 | XP_016872876.1 | |
XM_017017388.1 | 6825 | Silent Mutation | CTC,CTT | L,L 1825 | XP_016872877.1 | |
XM_017017389.1 | 6825 | Silent Mutation | CTC,CTT | L,L 1816 | XP_016872878.1 | |
XM_017017390.1 | 6825 | Silent Mutation | CTC,CTT | L,L 1255 | XP_016872879.1 |