Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCGTTGCTCATGTTCACCCGGCA[A/G]GCTGGACACTTCGTGGAGGGCTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605625 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYEOV PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYEOV - myeloma overexpressed | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293291.1 | 487 | Silent Mutation | CAA,CAG | Q,Q 63 | NP_001280220.1 | |
NM_001293294.1 | 487 | Silent Mutation | CAA,CAG | Q,Q 5 | NP_001280223.1 | |
NM_001293296.1 | 487 | Silent Mutation | CAA,CAG | Q,Q 5 | NP_001280225.1 | |
NM_001300923.1 | 487 | Silent Mutation | CAA,CAG | Q,Q 5 | NP_001287852.1 | |
NM_001300924.1 | 487 | Silent Mutation | CAA,CAG | Q,Q 5 | NP_001287853.1 | |
NM_138768.3 | 487 | Silent Mutation | CAA,CAG | Q,Q 63 | NP_620123.2 |