Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCAGGCCGACCGCTATGGATTCA[C/T]TGGGGGCAGCTCAGCAGAGCCAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176875 MIM: 603761 MIM: 610831 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1CA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP1CA - protein phosphatase 1 catalytic subunit alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAD9A - RAD9 checkpoint clamp component A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D10C - TBC1 domain family member 10C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256508.1 | 444 | Missense Mutation | ACT,ATT | T,I 43 | NP_001243437.1 | |
NM_198517.3 | 444 | Missense Mutation | ACT,ATT | T,I 43 | NP_940919.1 | |
XM_006718538.3 | 444 | Missense Mutation | ACT,ATT | T,I 43 | XP_006718601.1 | |
XM_006718539.3 | 444 | Missense Mutation | ACT,ATT | T,I 43 | XP_006718602.1 | |
XM_006718540.3 | 444 | Missense Mutation | ACT,ATT | T,I 43 | XP_006718603.1 | |
XM_006718541.3 | 444 | UTR 5 | XP_006718604.1 | |||
XM_006718542.3 | 444 | UTR 5 | XP_006718605.1 | |||
XM_006718543.2 | 444 | UTR 5 | XP_006718606.1 | |||
XM_006718544.3 | 444 | UTR 5 | XP_006718607.1 | |||
XM_011545001.2 | 444 | UTR 5 | XP_011543303.1 | |||
XM_011545002.2 | 444 | Missense Mutation | ACT,ATT | T,I 43 | XP_011543304.1 |