Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCACTAGTTTACAAGTATGTTGCC[A/G]CAGTCAGCTGATACCATTTAACCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603601 | ||||||||||||||||||||
Literature Links: |
PIK3C2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PIK3C2A - phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321378.1 | 5275 | Missense Mutation | GCG,GTG | A,V 1682 | NP_001308307.1 | |
NM_001321380.1 | 5275 | Missense Mutation | GCG,GTG | A,V 1302 | NP_001308309.1 | |
NM_002645.3 | 5275 | Missense Mutation | GCG,GTG | A,V 1682 | NP_002636.2 | |
XM_011520186.2 | 5275 | Missense Mutation | GCG,GTG | A,V 1626 | XP_011518488.1 | |
XM_017017924.1 | 5275 | Missense Mutation | GCG,GTG | A,V 1682 | XP_016873413.1 |