Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCTCGCCCGGAGAGTACACCCAC[C/T]GCGATTACGGCCACTCCAGTGTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609665 MIM: 605444 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NLRP14 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NLRP14 - NLR family pyrin domain containing 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_176822.3 | 908 | Intron | NP_789792.1 | |||
XM_011520044.1 | 908 | Intron | XP_011518346.1 |
RBMXL2 - RNA binding motif protein, X-linked like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014469.4 | 908 | Missense Mutation | CGC,TGC | R,C 241 | NP_055284.3 |