Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCGGTAAGGCACTGCGGCGGTGT[A/G]GCGCTCCCAGTCCCGGCCGTACTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602858 | ||||||||||||||||||||
Literature Links: |
DHCR7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DHCR7 - 7-dehydrocholesterol reductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163817.1 | 1634 | Missense Mutation | CAC,TAC | H,Y 462 | NP_001157289.1 | |
NM_001360.2 | 1634 | Missense Mutation | CAC,TAC | H,Y 462 | NP_001351.2 | |
XM_011544777.1 | 1634 | UTR 3 | XP_011543079.1 |