Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAGAAGCCAACAACTCCAGTGAA[C/T]GCTATAGAAATATTTTTCAGGACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613760 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC36A4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC36A4 - solute carrier family 36 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286139.1 | 1514 | Silent Mutation | GCA,GCG | A,A 322 | NP_001273068.1 | |
NM_152313.3 | 1514 | Silent Mutation | GCA,GCG | A,A 457 | NP_689526.2 | |
XM_017017174.1 | 1514 | UTR 3 | XP_016872663.1 | |||
XM_017017175.1 | 1514 | Silent Mutation | GCA,GCG | A,A 351 | XP_016872664.1 | |
XM_017017176.1 | 1514 | Silent Mutation | GCA,GCG | A,A 322 | XP_016872665.1 | |
XM_017017177.1 | 1514 | UTR 3 | XP_016872666.1 |