Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGGAACTCAATGTTGAAGGTGGT[A/G]TAGATGATGGGGTTCACGGCGCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 126450 | ||||||||||||||||||||
Literature Links: |
DRD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DRD2 - dopamine receptor D2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000795.3 | 1495 | Silent Mutation | TAC,TAT | Y,Y 426 | NP_000786.1 | |
NM_016574.3 | 1495 | Silent Mutation | TAC,TAT | Y,Y 397 | NP_057658.2 | |
XM_017017296.1 | 1495 | Silent Mutation | TAC,TAT | Y,Y 426 | XP_016872785.1 |