Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGGATGTGTTCTGGGCCAAGTA[C/T]ACAGGTGAGCACCGGGAAGGATTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176930 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
F2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
F2 - coagulation factor II, thrombin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000506.4 | 283 | Silent Mutation | TAC,TAT | Y,Y 87 | NP_000497.1 | |
NM_001311257.1 | 283 | Silent Mutation | TAC,TAT | Y,Y 71 | NP_001298186.1 |