Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAAGCGCCACTTCATCCAGGACA[C/G]CTGTCTCTATGAGTGCTCACCCAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 136425 MIM: 600829 | ||||||||||||||||||||
Literature Links: |
FOLR2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FOLR2 - folate receptor beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000803.4 | 450 | Missense Mutation | ACC,AGC | T,S 98 | NP_000794.3 | |
NM_001113534.1 | 450 | Missense Mutation | ACC,AGC | T,S 98 | NP_001107006.1 | |
NM_001113535.1 | 450 | Missense Mutation | ACC,AGC | T,S 98 | NP_001107007.1 | |
NM_001113536.1 | 450 | Missense Mutation | ACC,AGC | T,S 98 | NP_001107008.1 | |
XM_005273856.3 | 450 | Missense Mutation | ACC,AGC | T,S 107 | XP_005273913.1 |
INPPL1 - inositol polyphosphate phosphatase like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |