Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGACTGGGACATGGGGAACGAG[C/T]TCTTTCTGGCCTTCACCACATCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615755 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOXR1 - forkhead box R1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181721.2 | 238 | Missense Mutation | CTC,TTC | L,F 5 | NP_859072.1 | |
XM_017017575.1 | 238 | Missense Mutation | CTC,TTC | L,F 5 | XP_016873064.1 | |
XM_017017576.1 | 238 | Missense Mutation | CTC,TTC | L,F 5 | XP_016873065.1 |