Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAAACCCCTCCTTCTGCAGAGAG[C/T]GCACGTGGTCCTTGCAGAGCACAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603811 | ||||||||||||||||||||
Literature Links: |
BANF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BANF1 - barrier to autointegration factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EIF1AD - eukaryotic translation initiation factor 1A domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242481.1 | 425 | Missense Mutation | CAC,CGC | H,R 94 | NP_001229410.1 | |
NM_001242482.1 | 425 | Missense Mutation | CAC,CGC | H,R 94 | NP_001229411.1 | |
NM_001242483.1 | 425 | Missense Mutation | CAC,CGC | H,R 94 | NP_001229412.1 | |
NM_001242484.1 | 425 | Missense Mutation | CAC,CGC | H,R 94 | NP_001229413.1 | |
NM_001242485.1 | 425 | Missense Mutation | CAC,CGC | H,R 94 | NP_001229414.1 | |
NM_001242486.1 | 425 | Missense Mutation | CAC,CGC | H,R 94 | NP_001229415.1 | |
NM_032325.3 | 425 | Missense Mutation | CAC,CGC | H,R 94 | NP_115701.2 | |
XM_017018412.1 | 425 | Missense Mutation | CAC,CGC | H,R 94 | XP_016873901.1 |