Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTGCAACTAACCTTATCAATAGG[C/T]TCAATAAGAAAGTCATTGAACAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605861 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNPY2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNPY2 - canopy FGF signaling regulator 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PAN2 - PAN2 poly(A) specific ribonuclease subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127460.2 | 3074 | Silent Mutation | GAA,GAG | E,E 899 | NP_001120932.1 | |
NM_001166279.1 | 3074 | Silent Mutation | GAA,GAG | E,E 898 | NP_001159751.1 | |
NM_014871.4 | 3074 | Silent Mutation | GAA,GAG | E,E 895 | NP_055686.3 |