Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGCCAGTTCAAATTGGTCCTGCT[A/G]GGAGAATCTGCAGTGGGAAAGTCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179514 MIM: 606887 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB5B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAB5B - RAB5B, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252036.1 | 299 | Silent Mutation | CTA,CTG | L,L 26 | NP_001238965.1 | |
NM_001252037.1 | 299 | Silent Mutation | CTA,CTG | L,L 26 | NP_001238966.1 | |
NM_002868.3 | 299 | Silent Mutation | CTA,CTG | L,L 26 | NP_002859.1 | |
XM_005269051.2 | 299 | Silent Mutation | CTA,CTG | L,L 26 | XP_005269108.1 | |
XM_005269052.2 | 299 | Silent Mutation | CTA,CTG | L,L 26 | XP_005269109.1 |
SUOX - sulfite oxidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |