Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCGGCTCCGGCGTCAAACCCAA[A/G]CCACAAGTCTACTCTACCCTCCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603351 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OAS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OAS3 - 2'-5'-oligoadenylate synthetase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006187.3 | 508 | Silent Mutation | AAA,AAG | K,K 163 | NP_006178.2 | |
XM_005253889.3 | 508 | Silent Mutation | AAA,AAG | K,K 163 | XP_005253946.1 | |
XM_017019363.1 | 508 | Silent Mutation | AAA,AAG | K,K 163 | XP_016874852.1 |