Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCTCCGACAACAGGAATTTTCT[C/G]CGAGAGCGGGCCGGGCTCAGTTCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609858 | ||||||||||||||||||||
Literature Links: |
ETNK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ETNK1 - ethanolamine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039481.1 | 67 | Silent Mutation | CTC,CTG | L,L 16 | NP_001034570.1 | |
NM_018638.4 | 67 | Silent Mutation | CTC,CTG | L,L 16 | NP_061108.2 | |
XM_005253420.4 | 67 | Silent Mutation | CTC,CTG | L,L 16 | XP_005253477.1 | |
XM_017019580.1 | 67 | Silent Mutation | CTC,CTG | L,L 16 | XP_016875069.1 | |
XM_017019581.1 | 67 | Silent Mutation | CTC,CTG | L,L 16 | XP_016875070.1 |
LOC105369691 - uncharacterized LOC105369691 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |