Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTAAAGGTCCTCAGACTCCGGGA[C/T]TTTGGTGGATTTCTTGTGCAGAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609457 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMDHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMDHD1 - amidohydrolase domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HAL - histidine ammonia-lyase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258333.1 | 2250 | Missense Mutation | ATC,GTC | I,V 443 | NP_001245262.1 | |
NM_001258334.1 | 2250 | UTR 3 | NP_001245263.1 | |||
NM_002108.3 | 2250 | Missense Mutation | ATC,GTC | I,V 651 | NP_002099.1 | |
XM_011538249.2 | 2250 | Missense Mutation | ATC,GTC | I,V 367 | XP_011536551.1 | |
XM_017019246.1 | 2250 | Missense Mutation | ATC,GTC | I,V 341 | XP_016874735.1 |