Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAATCAACCACCGTATCTTTTCT[C/T]GAGTTCTTCGACAGAACTATCCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611477 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPAP3 - RNA polymerase II associated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146075.1 | 1992 | Missense Mutation | AAG,GAG | K,E 626 | NP_001139547.1 | |
NM_001146076.1 | 1992 | Missense Mutation | AAG,GAG | K,E 501 | NP_001139548.1 | |
NM_024604.2 | 1992 | Missense Mutation | AAG,GAG | K,E 660 | NP_078880.2 | |
XM_011538741.2 | 1992 | Missense Mutation | AAG,GAG | K,E 337 | XP_011537043.1 |