Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTCAGATTTACACCAAACCCTGA[A/G]TTTTCCACCTACCCACCAGCAGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609078 MIM: 608299 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDM2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KDM2B - lysine demethylase 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005366.1 | 268 | Intron | NP_001005366.1 | |||
NM_032590.4 | 268 | Intron | NP_115979.3 | |||
XM_005253955.3 | 268 | Intron | XP_005254012.1 | |||
XM_005253956.3 | 268 | Intron | XP_005254013.1 | |||
XM_005253961.4 | 268 | Intron | XP_005254018.1 | |||
XM_011538867.2 | 268 | Intron | XP_011537169.1 | |||
XM_011538868.2 | 268 | Intron | XP_011537170.1 | |||
XM_011538869.2 | 268 | Intron | XP_011537171.1 | |||
XM_011538875.2 | 268 | Intron | XP_011537177.1 |
RNF34 - ring finger protein 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256858.1 | 268 | Intron | NP_001243787.1 | |||
NM_025126.3 | 268 | Silent Mutation | GAA,GAG | E,E 46 | NP_079402.2 | |
NM_194271.2 | 268 | Silent Mutation | GAA,GAG | E,E 47 | NP_919247.1 |