Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTCCCAGAAGGGCACGAAGGTCT[C/T]CTGCAGCTGCCAGAAGGGATACAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611076 MIM: 608561 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NT5DC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NT5DC3 - 5'-nucleotidase domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031701.2 | 6547 | Intron | NP_001026871.1 | |||
XM_011538476.2 | 6547 | Intron | XP_011536778.1 | |||
XM_017019454.1 | 6547 | Intron | XP_016874943.1 |
STAB2 - stabilin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017564.9 | 6547 | Missense Mutation | TCC,TTC | S,F 2107 | NP_060034.9 | |
XM_011538537.2 | 6547 | Missense Mutation | TCC,TTC | S,F 2107 | XP_011536839.1 | |
XM_011538538.2 | 6547 | Missense Mutation | TCC,TTC | S,F 2107 | XP_011536840.1 | |
XM_011538539.2 | 6547 | Missense Mutation | TCC,TTC | S,F 1550 | XP_011536841.1 | |
XM_011538541.2 | 6547 | Missense Mutation | TCC,TTC | S,F 1240 | XP_011536843.1 | |
XM_011538542.2 | 6547 | Missense Mutation | TCC,TTC | S,F 794 | XP_011536844.1 | |
XM_017019585.1 | 6547 | Missense Mutation | TCC,TTC | S,F 2107 | XP_016875074.1 |