Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCACCTCCTGGGCTCCTCAGAGCC[A/G]GCTCATCCATCAGCTTGAGCTTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610109 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANO2 - anoctamin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278596.1 | 3208 | Missense Mutation | CCG,CTG | P,L 964 | NP_001265525.1 | |
NM_001278597.1 | 3208 | Missense Mutation | CCG,CTG | P,L 960 | NP_001265526.1 | |
XM_006718988.3 | 3208 | Missense Mutation | CCG,CTG | P,L 959 | XP_006719051.1 | |
XM_011520975.2 | 3208 | Missense Mutation | CCG,CTG | P,L 955 | XP_011519277.1 | |
XM_011520978.2 | 3208 | Missense Mutation | CCG,CTG | P,L 748 | XP_011519280.1 | |
XM_017019672.1 | 3208 | Intron | XP_016875161.1 |