Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCAGAATGGCCTCCTTTTCCATT[C/T]CAGAAATAGGAAGAAATTTAGACAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605545 | ||||||||||||||||||||
Literature Links: |
CD163 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD163 - CD163 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004244.5 | 3438 | Missense Mutation | GAA,GGA | E,G 1139 | NP_004235.4 | |
NM_203416.3 | 3438 | Intron | NP_981961.2 | |||
XM_005253528.4 | 3438 | Intron | XP_005253585.1 | |||
XM_005253529.4 | 3438 | Missense Mutation | GAA,GGA | E,G 1107 | XP_005253586.1 | |
XM_017020231.1 | 3438 | Intron | XP_016875720.1 |