Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGAGCTGCGTCGCCTCGTCTTG[A/C]CCTGCACCATCCTCCGGCGACGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612664 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RERG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RERG - RAS like estrogen regulated growth inhibitor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190726.1 | 864 | Missense Mutation | GGC,GTC | G,V 156 | NP_001177655.1 | |
NM_032918.2 | 864 | Missense Mutation | GGC,GTC | G,V 175 | NP_116307.1 | |
XM_017020121.1 | 864 | Missense Mutation | GGC,GTC | G,V 172 | XP_016875610.1 |