Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCTCGTGGGGCCGCCGCAGGCCC[C/G]CGTAGAAAGCAATGCGAGGCACGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615229 MIM: 603872 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1QL4 - complement C1q like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008223.1 | 1052 | Missense Mutation | GCG,GGG | A,G 114 | NP_001008224.1 | |
XM_011538270.2 | 1052 | Missense Mutation | GCG,GGG | A,G 114 | XP_011536572.1 |
TROAP - trophinin associated protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |