Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTCTGGTTGTCCATTGGAGACAT[C/T]GGGTTTGGCACCCTTGAGGACAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 154050 MIM: 603887 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIP - major intrinsic protein of lens fiber | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012064.3 | 506 | Missense Mutation | AAT,GAT | N,D 243 | NP_036196.1 | |
XM_011538354.1 | 506 | Missense Mutation | AAT,GAT | N,D 148 | XP_011536656.1 | |
XM_017019306.1 | 506 | Missense Mutation | AAT,GAT | N,D 124 | XP_016874795.1 |
TIMELESS - timeless circadian clock | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |