Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCACTGTTTATGCTGCACTTTG[C/T]TGTGAAATCAAGAAATTAAAATATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615748 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA1033 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA1033 - KIAA1033 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001293640.1 | 381 | Silent Mutation | TGC,TGT | C,C 98 | NP_001280569.1 | |
NM_015275.2 | 381 | Silent Mutation | TGC,TGT | C,C 98 | NP_056090.1 | |
XM_011538073.2 | 381 | Silent Mutation | TGC,TGT | C,C 98 | XP_011536375.1 | |
XM_011538074.2 | 381 | UTR 5 | XP_011536376.1 | |||
XM_011538077.2 | 381 | Silent Mutation | TGC,TGT | C,C 98 | XP_011536379.1 | |
XM_017019082.1 | 381 | Silent Mutation | TGC,TGT | C,C 98 | XP_016874571.1 |
LOC414300 - uncharacterized LOC414300 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |