Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAAAGAACCCTGGCTGGGGAAAA[A/G]GGAAGTGAAAAGAGATCTGTTTTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604082 | ||||||||||||||||||||
Literature Links: |
ZNF140 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF140 - zinc finger protein 140 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300776.1 | 1124 | UTR 5 | NP_001287705.1 | |||
NM_001300777.1 | 1124 | UTR 5 | NP_001287706.1 | |||
NM_001300778.1 | 1124 | UTR 5 | NP_001287707.1 | |||
NM_003440.3 | 1124 | Missense Mutation | AAG,AGG | K,R 69 | NP_003431.2 | |
XM_005266188.4 | 1124 | Missense Mutation | AAG,AGG | K,R 35 | XP_005266245.1 | |
XM_011534833.2 | 1124 | Missense Mutation | AAG,AGG | K,R 70 | XP_011533135.1 | |
XM_011534834.2 | 1124 | Missense Mutation | AAG,AGG | K,R 70 | XP_011533136.1 | |
XM_011534835.2 | 1124 | Missense Mutation | AAG,AGG | K,R 69 | XP_011533137.1 | |
XM_011534836.2 | 1124 | Missense Mutation | AAG,AGG | K,R 36 | XP_011533138.1 | |
XM_011534837.2 | 1124 | Missense Mutation | AAG,AGG | K,R 36 | XP_011533139.1 | |
XM_011534838.2 | 1124 | UTR 5 | XP_011533140.1 | |||
XM_011534839.2 | 1124 | UTR 5 | XP_011533141.1 | |||
XM_011534840.2 | 1124 | UTR 5 | XP_011533142.1 | |||
XM_017019924.1 | 1124 | UTR 5 | XP_016875413.1 | |||
XM_017019925.1 | 1124 | UTR 5 | XP_016875414.1 |