Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAATGAAGATGGCCCGGCCCAGCC[A/G]CTCCCTCAACGCCGCCCCCACTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103190 MIM: 601906 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARF3 - ADP ribosylation factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WNT10B - Wnt family member 10B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003394.3 | 674 | Missense Mutation | CGG,TGG | R,W 278 | NP_003385.2 | |
XM_011538722.1 | 674 | Missense Mutation | CGG,TGG | R,W 156 | XP_011537024.1 | |
XM_011538724.1 | 674 | UTR 3 | XP_011537026.1 | |||
XM_017019919.1 | 674 | Missense Mutation | CGG,TGG | R,W 156 | XP_016875408.1 |