Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGATCGGACTCTGGTCATCAAAGAA[G/T]TATCCAGTGAGGACATTGCTGACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613142 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DTX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DTX3 - deltex E3 ubiquitin ligase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIP4K2C - phosphatidylinositol-5-phosphate 4-kinase type 2 gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146258.1 | 584 | Missense Mutation | GTA,TTA | V,L 154 | NP_001139730.1 | |
NM_001146259.1 | 584 | Missense Mutation | GTA,TTA | V,L 136 | NP_001139731.1 | |
NM_001146260.1 | 584 | Intron | NP_001139732.1 | |||
NM_024779.4 | 584 | Missense Mutation | GTA,TTA | V,L 154 | NP_079055.3 | |
XM_005269152.2 | 584 | Missense Mutation | GTA,TTA | V,L 154 | XP_005269209.1 | |
XM_011538747.1 | 584 | Missense Mutation | GTA,TTA | V,L 154 | XP_011537049.1 | |
XM_017019973.1 | 584 | UTR 5 | XP_016875462.1 |