Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCTGAGAAGCCGCTCCCGTTCA[C/T]TGGTCAGCCGCTCCAGCTCCCGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164017 MIM: 601490 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HNRNPA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HNRNPA1 - heterogeneous nuclear ribonucleoprotein A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NFE2 - nuclear factor, erythroid 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136023.2 | 1331 | Missense Mutation | AAT,AGT | N,S 303 | NP_001129495.1 | |
NM_001261461.1 | 1331 | Missense Mutation | AAT,AGT | N,S 303 | NP_001248390.1 | |
NM_006163.2 | 1331 | Missense Mutation | AAT,AGT | N,S 303 | NP_006154.1 | |
XM_005268906.4 | 1331 | Missense Mutation | AAT,AGT | N,S 303 | XP_005268963.1 |