Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCTGCGCCGAAAGTTCCGTGCAC[A/C]CTGGCGATCGTGTCCCGTACGAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609992 MIM: 615998 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POP5 - POP5 homolog, ribonuclease P/MRP subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015918.3 | 149 | Missense Mutation | AGG,AGT | R,S 39 | NP_057002.2 | |
NM_198202.1 | 149 | Missense Mutation | AGG,AGT | R,S 39 | NP_937845.1 | |
XM_011538441.1 | 149 | Missense Mutation | AGG,AGT | R,S 39 | XP_011536743.1 |
RNF10 - ring finger protein 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |