Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTTACCTGCAGTCTCACACTTAC[C/T]AACAATTTTCCACTTAGGAATCCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610642 | ||||||||||||||||||||
Literature Links: |
ERP27 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ERP27 - endoplasmic reticulum protein 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300784.1 | 815 | Silent Mutation | TTA,TTG | L,L 157 | NP_001287713.1 | |
NM_152321.3 | 815 | Silent Mutation | TTA,TTG | L,L 258 | NP_689534.1 |