Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCCCGTACTCAGGCTTGGGCTCC[C/T]GCCAGCTCTTGGATGGCAGCTCCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615104 | ||||||||||||||||||||
Literature Links: |
NCKAP5L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NCKAP5L - NCK associated protein 5 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037806.3 | 3628 | Missense Mutation | CAG,CGG | Q,R 1044 | NP_001032895.2 | |
XM_005269050.3 | 3628 | Missense Mutation | CAG,CGG | Q,R 1044 | XP_005269107.1 | |
XM_006719525.2 | 3628 | Missense Mutation | CAG,CGG | Q,R 1044 | XP_006719588.1 | |
XM_011538610.2 | 3628 | Missense Mutation | CAG,CGG | Q,R 1044 | XP_011536912.1 | |
XM_011538611.2 | 3628 | Missense Mutation | CAG,CGG | Q,R 1044 | XP_011536913.1 |