Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCACCTGCACTCCTCGCTCTCC[A/G]GCAGCTTGCGGTAGGTGGCGATCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608246 | ||||||||||||||||||||
Literature Links: |
KRT72 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KRT72 - keratin 72 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146225.1 | 1080 | Missense Mutation | CCG,CTG | P,L 431 | NP_001139697.1 | |
NM_001146226.1 | 1080 | Missense Mutation | CCG,CTG | P,L 389 | NP_001139698.1 | |
NM_080747.2 | 1080 | Missense Mutation | CCG,CTG | P,L 431 | NP_542785.1 | |
XM_017018833.1 | 1080 | Missense Mutation | CCG,CTG | P,L 365 | XP_016874322.1 | |
XM_017018834.1 | 1080 | Missense Mutation | CCG,CTG | P,L 362 | XP_016874323.1 | |
XM_017018835.1 | 1080 | Missense Mutation | CCG,CTG | P,L 359 | XP_016874324.1 | |
XM_017018836.1 | 1080 | Missense Mutation | CCG,CTG | P,L 339 | XP_016874325.1 | |
XM_017018837.1 | 1080 | Missense Mutation | CCG,CTG | P,L 191 | XP_016874326.1 |