Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTGTTGGGAGAGGGTGGAGGACT[C/G]GAGTTACGTTTTGTAGGAGCATCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 108731 | ||||||||||||||||||||
Literature Links: |
ATP2B1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP2B1 - ATPase plasma membrane Ca2+ transporting 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001323.1 | 3974 | UTR 3 | NP_001001323.1 | |||
NM_001682.2 | 3974 | Silent Mutation | TCC,TCG | S,S 1177 | NP_001673.2 | |
XM_011538407.2 | 3974 | Silent Mutation | TCC,TCG | S,S 1206 | XP_011536709.1 | |
XM_017019357.1 | 3974 | Silent Mutation | TCC,TCG | S,S 1170 | XP_016874846.1 | |
XM_017019358.1 | 3974 | Silent Mutation | TCC,TCG | S,S 1141 | XP_016874847.1 | |
XM_017019359.1 | 3974 | UTR 3 | XP_016874848.1 | |||
XM_017019360.1 | 3974 | Intron | XP_016874849.1 |