Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCCTGGCATTCTTGAAAATATT[C/T]GTCAAACATAGAGGAAGACTCATCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604225 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARPC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARPC3 - actin related protein 2/3 complex subunit 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPN3 - GPN-loop GTPase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164372.1 | 1362 | Missense Mutation | AAA,GAA | K,E 315 | NP_001157844.1 | |
NM_001164373.1 | 1362 | Missense Mutation | AAA,GAA | K,E 286 | NP_001157845.1 | |
NM_016301.3 | 1362 | Missense Mutation | AAA,GAA | K,E 276 | NP_057385.3 | |
XM_005253896.4 | 1362 | Missense Mutation | AAA,GAA | K,E 449 | XP_005253953.3 | |
XM_017019394.1 | 1362 | Missense Mutation | AAA,GAA | K,E 254 | XP_016874883.1 |