Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCGGGAGCCACCTGAATGGCGA[C/T]CTGGATCCAGACGACAGGGAAGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601870 | ||||||||||||||||||||
Literature Links: |
METAP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
METAP2 - methionyl aminopeptidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317182.1 | 185 | Silent Mutation | GAC,GAT | D,D 17 | NP_001304111.1 | |
NM_001317183.1 | 185 | Silent Mutation | GAC,GAT | D,D 17 | NP_001304112.1 | |
NM_006838.3 | 185 | Silent Mutation | GAC,GAT | D,D 17 | NP_006829.1 | |
XM_005268583.3 | 185 | Silent Mutation | GAC,GAT | D,D 17 | XP_005268640.1 |