Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATATGTAGTTCCAAGGAGAAGTCA[C/G]AGTTCCACCTTTGGAGTCTTTCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610642 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERP27 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERP27 - endoplasmic reticulum protein 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300784.1 | 860 | Silent Mutation | CTC,CTG | L,L 172 | NP_001287713.1 | |
NM_152321.3 | 860 | Silent Mutation | CTC,CTG | L,L 273 | NP_689534.1 |