Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605476 MIM: 123829 MIM: 613336 MIM: 181035 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGAP2 - ArfGAP with GTPase domain, ankyrin repeat and PH domain 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CDK4 - cyclin dependent kinase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MARCH9 - membrane associated ring-CH-type finger 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6759 - microRNA 6759 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSPAN31 - tetraspanin 31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005981.3 | 1148 | Silent Mutation | GTA,GTG | V,V 23 | NP_005972.1 | |
XM_005269074.3 | 1148 | Silent Mutation | GTA,GTG | V,V 113 | XP_005269131.2 | |
XM_017019793.1 | 1148 | Intron | XP_016875282.1 |