Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAACACCATCTGTGACGTCCTGC[A/G]GGAACCCGAACAGTTCCCCCTGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603350 MIM: 603351 | ||||||||||||||||||||
Literature Links: |
OAS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OAS2 - 2'-5'-oligoadenylate synthetase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032731.1 | 277 | Missense Mutation | CAG,CGG | Q,R 46 | NP_001027903.1 | |
NM_002535.2 | 277 | Missense Mutation | CAG,CGG | Q,R 46 | NP_002526.2 | |
NM_016817.2 | 277 | Missense Mutation | CAG,CGG | Q,R 46 | NP_058197.2 |
OAS3 - 2'-5'-oligoadenylate synthetase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |