Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACAGTTTTCTCACTTTCGCCATC[A/G]GGCACTGGAGCATCTGGCCTCCTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC53 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC53 - coiled-coil domain containing 53 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301107.1 | 684 | Silent Mutation | CCC,CCT | P,P 173 | NP_001288036.1 | |
NM_016053.3 | 684 | Silent Mutation | CCC,CCT | P,P 174 | NP_057137.1 | |
XM_011538423.1 | 684 | Intron | XP_011536725.1 | |||
XM_017019383.1 | 684 | Intron | XP_016874872.1 |