Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGAAGAATTCTGTAACTTAATA[G/T]GAGAGGATTGCCTGGGTCTGTTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603141 MIM: 610848 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPFIBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPFIBP1 - PPFIA binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
REP15 - RAB15 effector protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029874.1 | 534 | Missense Mutation | GGA,TGA | G,* 156 | NP_001025045.1 |